siRNA Details
Gene Symbol: Ube2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026785.2 | NP_081061.1 | 3 | 205 | ||
GenBank mRNA | AK003722.1 | - | 3 | 206 | ||
AK136186.1 | - | 3 | 206 | |||
AK160740.1 | - | 3 | 237 | |||
BC147032.1 | - | 3 | 186 | 9056036 | ||
BC147033.1 | - | 3 | 186 | 9056038 | ||
BC171983.1 | - | 3 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
