siRNA Details
Gene Symbol: Nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040395.4 | NP_001035485.2 | 10 | 1261 | ||
NM_001085410.2 | NP_001078879.1 | 11 | 1327 | |||
NM_001286253.1 | NP_001273182.1 | 10 | 1261 | |||
NM_001286255.1 | NP_001273184.1 | 9 | 1177 | |||
GenBank mRNA | AK003858.1 | - | 10 | 838 | ||
AK016988.1 | - | 10 | 827 | |||
AK046666.1 | - | 11 | 1328 | |||
AK078318.1 | - | 11 | 930 | |||
AK149267.1 | - | 9 | 757 | |||
AK162601.1 | - | 10 | 835 | |||
BC048411.1 | - | 1 | 57 | |||
BC115776.1 | - | 10 | 840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
