siRNA Details
Gene Symbol: Fam198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133187.3 | NP_573450.2 | 3, 4 | 1642 | ||
GenBank mRNA | AF285091.1 | - | 3, 4 | 1584 | ||
AK052367.1 | - | 3, 4 | 609 | |||
AK143522.1 | - | 3, 4 | 1373 | |||
AK195202.1 | - | 1, 2 | 120 | |||
BC056975.1 | - | 3, 4 | 1411 | 6827227 | ||
EU797522.1 | - | 3, 4 | 1420 |
