siRNA Details
Gene Symbol: MEP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005588.2 | NP_005579.2 | 10 | 1150 | ||
XM_011514628.1 | XP_011512930.1 | 9 | 1207 | |||
XM_011514629.1 | XP_011512931.1 | 10 | 1150 | |||
GenBank mRNA | AF478685.1 | - | 10 | 1118 | ||
AK225212.1 | - | 10 | 1140 | |||
AK290282.1 | - | 10 | 1118 | |||
AK315246.1 | - | 10 | 1122 | |||
AK315840.1 | - | 10 | 1120 | |||
BC131714.1 | - | 10 | 1121 | 40122874 | ||
BC143650.1 | - | 8 | 1289 | |||
BC143651.1 | - | 9 | 1200 | 9052165 | ||
M82962.1 | - | 10 | 1117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
