siRNA Details
Gene Symbol: Kansl1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170981.1 | - | 2 | 945 | ||
AK172645.1 | - | 2 | 869 | |||
AK220534.1 | - | 2 | 867 | |||
BC141036.1 | - | 3 | 826 | 9055648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
