siRNA Details
Gene Symbol: Hddc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026812.2 | NP_081088.1 | 1 | 45 | ||
GenBank mRNA | AK004078.1 | - | 1 | 46 | ||
BC038310.1 | - | 1 | 58 | 5119020 | ||
BC055847.1 | - | 1 | 58 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
