siRNA Details
Gene Symbol: Hddc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026812.2 | NP_081088.1 | 1, 2 | 121 | ||
GenBank mRNA | AK004078.1 | - | 1, 2 | 122 | ||
BC038310.1 | - | 1, 2 | 134 | 5119020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
