siRNA Details
Gene Symbol: Rabl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026817.3 | NP_081093.2 | 7, 8 | 653 | ||
GenBank mRNA | AK170205.1 | - | 4, 5 | 341 | ||
AK210352.1 | - | 2, 3 | 132 | |||
BC030376.1 | - | 7, 8 | 620 | 5374611 | ||
BQ887131.1 | - | 7, 8 | 645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
