siRNA Details
Gene Symbol: 1110032F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167996.1 | NP_001161468.1 | 1 | 1776 | ||
GenBank mRNA | AK004029.1 | - | 1 | 49 | ||
AK082199.1 | - | 1 | 1521 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006232486.
Gene Ontology Categories:
