siRNA Details
Gene Symbol: Rbfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199197.1 | NP_954667.1 | 2 | 372 | ||
GenBank mRNA | AK004019.1 | - | 2 | 391 | ||
AK145807.1 | - | 2 | 411 | |||
BC064066.1 | - | 2 | 372 | 935634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
