siRNA Details
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037742.2 | NP_001032831.2 | 8 | 1083 | ||
GenBank mRNA | BC023868.1 | - | 8 | 1075 | ||
BC058590.1 | - | 7 | 1399 | |||
BC099495.1 | - | 8 | 825 | |||
BC132289.1 | - | 8 | 932 | 40130566 | ||
BC144765.1 | - | 8 | 898 | |||
BC144766.1 | - | 8 | 913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
