siRNA Details
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037742.2 | NP_001032831.2 | 8 | 977 | ||
GenBank mRNA | BC023868.1 | - | 8 | 969 | ||
BC058590.1 | - | 7 | 1293 | |||
BC099495.1 | - | 8 | 719 | |||
BC132289.1 | - | 8 | 826 | 40130566 | ||
BC144765.1 | - | 8 | 792 | |||
BC144766.1 | - | 8 | 807 |

More Information
Other Species Targeted

Gene Ontology Categories:
