siRNA Details
Gene Symbol: Tmem219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026827.1 | NP_081103.1 | 3 | 427 | ||
NM_028389.1 | NP_082665.1 | 3 | 424 | |||
GenBank mRNA | AK004043.1 | - | 2 | 387 | ||
AK013646.1 | - | 2 | 229 | |||
AK152526.1 | - | 3 | 389 | |||
BC023442.1 | - | 3 | 402 | 5026775 | ||
BC046763.1 | - | 2 | 569 | 5693779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
