siRNA Details
Gene Symbol: Zfp740 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289690.1 | NP_001276619.1 | 4 | 822 | ||
NM_001289691.1 | NP_001276620.1 | 6 | 1021 | |||
NM_001289695.1 | NP_001276624.1 | 5 | 696 | |||
GenBank mRNA | AK144707.1 | - | 6 | 1018 | ||
BC030410.1 | - | 1 | 527 | 5367159 | ||
BC066011.1 | - | 5 | 590 | 6418561 | ||
CA319591.1 | - | 4 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
