siRNA Details
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083895.3 | NP_001077364.2 | 8, 9 | 1457 | ||
NM_026838.4 | NP_081114.2 | 9, 10 | 1509 | |||
GenBank mRNA | AF393640.1 | - | 7, 8 | 1027 | ||
AK136935.1 | - | 8, 9 | 1312 | |||
BC028307.1 | - | 9, 10 | 1238 | 3488526 | ||
BQ560499.1 | - | 1, 2 | 32 |

More Information
Other Species Targeted

Gene Ontology Categories:
