siRNA Details
Gene Symbol: ABL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136000.2 | NP_001129472.1 | 13 | 3188 | ||
NM_001168236.1 | NP_001161708.1 | 11 | 3721 | |||
NM_001168237.1 | NP_001161709.1 | 13 | 3475 | |||
NM_001168238.1 | NP_001161710.1 | 12 | 3412 | |||
NM_001168239.1 | NP_001161711.1 | 12 | 3125 | |||
NM_005158.4 | NP_005149.4 | 12 | 3497 | |||
NM_007314.3 | NP_009298.1 | 12 | 3784 | |||
XM_005245088.2 | XP_005245145.1 | 11 | 3419 | |||
XM_017001035.1 | XP_016856524.1 | 11 | 6094 | |||
GenBank mRNA | BC065912.1 | - | 12 | 3464 | 5173213 | |
BX538317.1 | - | 13 | 3227 | |||
FJ542283.1 | - | 12 | 3080 | |||
FJ542284.1 | - | 13 | 3188 | |||
FJ542285.1 | - | 11 | 3434 | |||
FJ542286.1 | - | 12 | 3125 | |||
M35296.1 | - | 12 | 3701 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
