siRNA Details
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134255.3 | NP_599016.2 | 7 | 887 | ||
GenBank mRNA | AK028761.1 | - | 7 | 827 | ||
AK031743.1 | - | 7 | 890 | |||
AK045274.1 | - | 7 | 866 | |||
AK051580.1 | - | 7 | 884 | |||
AK085663.1 | - | 7 | 850 | |||
AK085696.1 | - | 7 | 837 | |||
AK088153.1 | - | 7 | 825 | |||
AK162451.1 | - | 7 | 888 | |||
AK167755.1 | - | 7 | 827 | |||
AK168360.1 | - | 7 | 827 | |||
AK168987.1 | - | 7 | 814 | |||
BC022911.1 | - | 7 | 796 | 4457285 | ||
KF987081.1 | - | 7 | 770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
