siRNA Details
Gene Symbol: Mrpl52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026851.2 | NP_081127.1 | 5 | 338 | ||
GenBank mRNA | AK004194.1 | - | 5 | 339 | ||
AK150806.1 | - | 5 | 338 | |||
AK151615.1 | - | 5 | 338 | |||
AK152876.1 | - | 5 | 325 | |||
AK164629.1 | - | 5 | 336 | |||
BC028522.1 | - | 5 | 371 | 1448458 | ||
BC060043.1 | - | 4 | 348 | 5683366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
