siRNA Details
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103181.1 | NP_001096651.1 | 7 | 1083 | ||
NM_054040.3 | NP_473381.1 | 7 | 1925 | |||
GenBank mRNA | AF219945.1 | - | 7 | 1329 | ||
AK147643.1 | - | 7 | 1023 | |||
AK173165.1 | - | 7 | 1925 | |||
BC138024.1 | - | 7 | 1260 | 8861043 | ||
BC138025.1 | - | 7 | 1260 | 8861044 |
