siRNA Details
Gene Symbol: Tulp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103181.1 | NP_001096651.1 | 5 | 723 | ||
NM_054040.3 | NP_473381.1 | 5 | 1565 | |||
GenBank mRNA | AF219945.1 | - | 5 | 969 | ||
AK147643.1 | - | 5 | 663 | |||
AK173165.1 | - | 5 | 1565 | |||
BC138024.1 | - | 5 | 900 | 8861043 | ||
BC138025.1 | - | 5 | 900 | 8861044 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109137, XM_006227887, XM_008758736, XM_008758737.
Gene Ontology Categories:
