siRNA Details
Gene Symbol: Rnf208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176834.2 | NP_789804.2 | 1 | 900 | ||
GenBank mRNA | AK004344.1 | - | 1 | 124 | ||
BC030073.1 | - | 2 | 702 | 5254639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
