siRNA Details
Gene Symbol: Lrrn4cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013019.2 | NP_001013037.1 | 2 | 473 | ||
GenBank mRNA | AK087615.1 | - | 474 | |||
AK158268.1 | - | 2 | 411 | |||
BC107244.2 | - | 2 | 446 | 40052213 | ||
BC107245.2 | - | 2 | 446 | 40052216 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
