siRNA Details
Gene Symbol: Rnf122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175136.2 | NP_780345.1 | 3, 4 | 537 | ||
GenBank mRNA | AK077856.1 | - | 3, 4 | 557 | ||
AK085376.1 | - | 3, 4 | 519 | |||
AK136206.1 | - | 3, 4 | 555 | |||
AK171823.1 | - | 3, 4 | 557 | |||
BC038399.1 | - | 3, 4 | 537 | 4947596 | ||
BC050268.1 | - | 2, 3 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
