siRNA Details
Gene Symbol: Rnf122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175136.2 | NP_780345.1 | 6 | 1557 | ||
GenBank mRNA | AK004356.1 | - | 1 | 776 | ||
AK028959.1 | - | 1 | 81 | |||
AK046909.1 | - | 1 | 94 | |||
AK085376.1 | - | 6 | 1541 | |||
AK136206.1 | - | 6 | 1579 | |||
AK171823.1 | - | 6 | 1581 | |||
BC038399.1 | - | 6 | 1559 | 4947596 | ||
BC050268.1 | - | 5 | 1246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
