siRNA Details
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133701.2 | NP_598462.1 | 2, 3 | 377 | ||
GenBank mRNA | AK011639.1 | - | 2, 3 | 319 | ||
AK081998.1 | - | 2, 3 | 312 | |||
AK145461.1 | - | 2, 3 | 380 | |||
AK166890.1 | - | 2, 3 | 329 | |||
BC014869.1 | - | 2, 3 | 304 | 3602158 | ||
BC023691.1 | - | 2, 3 | 306 | 5344332 | ||
BC025030.1 | - | 2, 3 | 323 | 4947416 | ||
BY735790.1 | - | 2, 3 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
