siRNA Details
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026880.2 | NP_081156.2 | 4 | 1016 | ||
GenBank mRNA | AB053476.1 | - | 4 | 948 | ||
AK050322.1 | - | 4 | 996 | |||
AK155474.1 | - | 4 | 1002 | |||
AK165480.1 | - | 4 | 995 | |||
AK180089.1 | - | 1 | 77 | |||
AK198589.1 | - | 1 | 78 | |||
BC018594.1 | - | 4 | 979 | |||
BC067066.1 | - | 4 | 984 | 30542123 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
