siRNA Details
Gene Symbol: Eif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026892.3 | NP_081168.1 | 2 | 363 | ||
GenBank mRNA | AI154212.1 | - | 2 | 365 | ||
AK005192.1 | - | 2 | 330 | |||
AK013965.1 | - | 2 | 336 | |||
BC030319.1 | - | 2 | 232 | 5372299 | ||
BC033505.1 | - | 2 | 334 | 4209443 |

More Information
Other Species Targeted

Gene Ontology Categories:
