siRNA Details
Gene Symbol: Dcaf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026893.3 | NP_081169.2 | 2 | 627 | ||
GenBank mRNA | AK005099.1 | - | 2 | 573 | ||
AK030173.1 | - | 2 | 579 | |||
AK031089.1 | - | 2 | 557 | |||
AK083369.1 | - | 1 | 1577 | |||
AK087517.1 | - | 2 | 510 | |||
AK090028.1 | - | 2 | 576 | |||
AK143635.1 | - | 2 | 580 | |||
AK173289.1 | - | 2 | 487 | |||
AK188223.1 | - | 1 | 135 | |||
BC023852.1 | - | 2 | 563 | 5346910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
