siRNA Details
Gene Symbol: Med27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026896.5 | NP_081172.2 | 2 | 361 | ||
NM_029615.1 | NP_083891.1 | 2 | 361 | |||
GenBank mRNA | AK005251.1 | - | 2 | 336 | ||
AK009764.1 | - | 2 | 342 | |||
AK147181.1 | - | 2 | 358 | |||
BC016537.1 | - | 2 | 352 | 3585027 | ||
CJ072886.1 | - | 2 | 362 |
