siRNA Details
Gene Symbol: Zfp580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026900.1 | NP_081176.1 | 2 | 831 | ||
GenBank mRNA | AK005257.1 | - | 2 | 826 | ||
BC066001.1 | - | 2 | 779 | 5700640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
