siRNA Details
Gene Symbol: Snx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026912.1 | NP_081188.1 | 1 | 156 | ||
GenBank mRNA | BC016091.1 | - | 1 | 156 | 4503873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
