siRNA Details
Gene Symbol: Zg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026918.2 | NP_081194.1 | 2 | 177 | ||
GenBank mRNA | BC031800.1 | - | 2 | 177 | 4208847 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
