siRNA Details
Gene Symbol: Cml5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023493.2 | NP_075982.2 | 3 | 821 | ||
GenBank mRNA | AF187099.1 | - | 1 | 306 | ||
AK007530.1 | - | 3 | 821 | |||
BC024605.1 | - | 3 | 804 | 4236653 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
