siRNA Details
Gene Symbol: 1810013L24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081400.3 | NP_001074869.1 | 3 | 910 | ||
GenBank mRNA | AK007485.1 | - | 3 | 910 | ||
BC038008.1 | - | 2 | 347 | |||
BC116778.1 | - | 3 | 893 | 40126097 | ||
BC116780.1 | - | 3 | 893 | 40126099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
