siRNA Details
Gene Symbol: 1810011H11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163616.1 | NP_001157088.1 | 3 | 269 | ||
GenBank mRNA | AK007434.1 | - | 3 | 270 | ||
AK014636.1 | - | 2 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
