siRNA Details
Gene Symbol: Tmem97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133706.2 | NP_598467.1 | 1 | 164 | ||
GenBank mRNA | AK155107.1 | - | 1 | 176 | ||
AK171954.1 | - | 1 | 143 | |||
BC018156.1 | - | 1 | 164 | 4192256 | ||
BC093511.1 | - | 1 | 141 | 5124076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
