siRNA Details
Gene Symbol: Sult1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026935.4 | NP_081211.3 | 8 | 1030 | ||
GenBank mRNA | AK007384.1 | - | 8 | 890 | ||
AK144336.1 | - | 8 | 970 | |||
AY005469.1 | - | 8 | 1030 | |||
BC022665.1 | - | 8 | 1023 | 4223678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
