siRNA Details
Gene Symbol: Trim15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024134.2 | NP_001019305.1 | 5 | 849 | ||
NM_001177872.1 | NP_001171343.1 | 5 | 872 | |||
GenBank mRNA | BC027186.1 | - | 5 | 849 | 4215801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
