siRNA Details
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026940.4 | NP_081216.1 | 3 | 358 | ||
GenBank mRNA | AK007503.1 | - | 3 | 347 | ||
AK032396.1 | - | 388 | ||||
AK076869.1 | - | 968 | ||||
AK140044.1 | - | 463 | ||||
AK141482.1 | - | 1 | 638 | |||
BC030425.1 | - | 3 | 360 | |||
BC115584.1 | - | 3 | 346 | 40102346 | ||
BC115585.1 | - | 3 | 346 | 40102347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
