siRNA Details
Gene Symbol: Alkbh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026944.1 | NP_081220.1 | 3 | 271 | ||
GenBank mRNA | AK145586.1 | - | 3 | 657 | ||
BC018196.1 | - | 3 | 271 | 3993214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
