siRNA Details
Gene Symbol: Cd209f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026956.2 | NP_081232.2 | 8 | 730 | ||
GenBank mRNA | AK007638.1 | - | 8 | 787 | ||
BC021323.1 | - | 8 | 811 | |||
BC038121.1 | - | 8 | 1133 | |||
BC046292.1 | - | 6 | 771 | |||
BC052782.1 | - | 8 | 721 | |||
BC099497.1 | - | 8 | 725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
