siRNA Details
Gene Symbol: Cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026977.2 | NP_081253.1 | 3 | 414 | ||
GenBank mRNA | AK029090.1 | - | 3 | 414 | ||
AK145962.1 | - | 4 | 661 | |||
AK159252.1 | - | 136 | ||||
AK169779.1 | - | 3 | 686 | |||
AK187356.1 | - | 2 | 34 | |||
BC013714.1 | - | 2 | 187 | |||
BC028815.1 | - | 3 | 399 | 5126467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
