siRNA Details
Gene Symbol: Erp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026983.2 | NP_081259.1 | 3, 4 | 396 | ||
GenBank mRNA | AK007684.1 | - | 3, 4 | 397 | ||
AK007800.1 | - | 3, 4 | 317 | |||
AK131830.1 | - | 3, 4 | 397 | |||
BC037376.1 | - | 2, 3 | 232 | |||
BC104406.1 | - | 3, 4 | 384 | 40050532 | ||
BC104407.2 | - | 3, 4 | 384 | 40050533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
