siRNA Details
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093752.1 | NP_001087221.1 | 12 | 1294 | ||
NM_001093753.1 | NP_001087222.1 | 11 | 1276 | |||
GenBank mRNA | AK081511.1 | - | 9 | 1101 | ||
AK137854.1 | - | 4 | 368 | |||
AK141191.1 | - | 5 | 406 | |||
AK146719.1 | - | 11 | 1248 | |||
AK183815.1 | - | 3 | 172 | |||
BC055763.1 | - | 13 | 1181 | |||
BC069945.1 | - | 11 | 1226 | |||
BC138145.1 | - | 9 | 1029 | 8861164 | ||
BC145162.1 | - | 8 | 888 | 9053713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
