siRNA Details
Gene Symbol: Snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029394.3 | NP_083670.1 | 2 | 212 | ||
GenBank mRNA | AK012711.1 | - | 2 | 104 | ||
AK017610.1 | - | 2 | 106 | |||
BC051168.1 | - | 2 | 215 | 6306904 |

More Information
Other Species Targeted

Gene Ontology Categories:
