siRNA Details
Gene Symbol: Qrich1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114119.1 | NP_001107591.1 | 7 | 2021 | ||
NM_175143.5 | NP_780352.2 | 6 | 1897 | |||
GenBank mRNA | AK083212.1 | - | 6 | 1882 | ||
AK151491.1 | - | 7 | 1995 | |||
AK151530.1 | - | 7 | 1995 | |||
BC006738.1 | - | 6 | 1874 | 3156574 | ||
BC120915.1 | - | 5 | 796 | |||
BC120916.1 | - | 6 | 1828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
