siRNA Details
Gene Symbol: Zfp688 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026999.4 | NP_081275.3 | 3 | 694 | ||
GenBank mRNA | AK013037.1 | - | 2 | 974 | ||
AK170857.1 | - | 2 | 2416 | |||
BC027683.1 | - | 2 | 977 | 1345974 | ||
BC038680.1 | - | 1 | 265 | |||
BC052512.1 | - | 3 | 534 | 1362502 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
