siRNA Details
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027009.2 | NP_081285.1 | 6, 7 | 772 | ||
GenBank mRNA | AK042393.1 | - | 6, 7 | 773 | ||
AK145126.1 | - | 6, 7 | 799 | |||
AK167047.1 | - | 6, 7 | 773 | |||
AK209135.1 | - | 3, 4 | 349 | |||
BC026795.1 | - | 6, 7 | 743 | 4015134 |
