siRNA Details
Gene Symbol: 1700001J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008547.1 | NP_001008547.1 | 3, 4 | 414 | ||
GenBank mRNA | AK005597.1 | - | 3, 4 | 410 | ||
BC048528.1 | - | 3, 4 | 414 | 6774214 |
