siRNA Details
Gene Symbol: Asb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027027.2 | NP_081303.1 | 573 | |||
GenBank mRNA | AF398970.1 | - | 4 | 505 | ||
AK005878.1 | - | 574 | ||||
BC061038.1 | - | 565 | 6703903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
